Crile Hemostatic Forceps, OR Grade, Sklar
Supplier: Sklar
17-3055 17-3062 17-3155 17-3162 17-3075 17-3168 17-3175 18-1952 18-2052
82030-012EA 102.24 CAD
82030-012 82030-014 82030-016 82030-018 10193-286 10194-784 10194-786 10194-832 10194-834
Crile Hemostatic Forceps, OR Grade, Sklar
Forceps
Crile Hemostatic Forceps, also known as a Crile Hemostat, are used for clamping blood vessels or tissue before cauterization or ligation. They may also be used for soft tissue dissection no deeper than 6 - 8" or in laparotomy procedures.
Horizontal serrations the entire length of the jaw, and jaws half the length of the shank, as well as a ratchet locking mechanism allow for a tight, secure hold on vessels.
Censitrac Ready products are industry standard and constructed of stainless steel.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.