Merit™ Hartmann Alligator Forceps, Physician Grade, Sklar
Supplier: Sklar
98-232 98-224 98-209
10192-332EA 193.18 CAD
10192-332 10192-446 10193-768
Merit™ Hartmann Alligator Forceps, Physician Grade, Sklar
Forceps
Merit™ Hartmann Alligator Forceps are an ENT instrument used in myringotomy procedures, to insert or remove tubes or dressings and packing.
The serrated tips securely hold and remove foreign objects, granulation tissue, and cerumen. Merit™ instruments are office-grade products used for routine and in-office procedures.
These high quality instruments are crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.