Utility Scissors, Mid-Grade, Sklar
Supplier: Sklar
21-101 21-105 95-105
89023-024PK 264.5 CAD
89023-024 89023-032 89023-446
Utility Scissors, Mid-Grade, Sklar
Scissors
Surgi-OR™ Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes.
  • Cost-effective
  • Reliable instrumentation
  • For use in operating room and surgical centers
The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages.
These scissors feature plastic finger ring handles with one ring that is larger than the other.
Caution: They should never be used to cut tissue or sutures.