Micro Scissors, Sklar
Supplier: Sklar
98-7892 98-7813 98-7814 98-7837
10185-190EA 2904.07 CAD
10185-190 75996-306 75996-308 75996-324
Micro Scissors, Sklar
Scissors
Sklar's® micro scissors are used for the dissection of tissues, blood vessels and nerves.
  • Reusable
  • Nonsterile
Available with a flat or round handle. The straight scissors may be used to cut sutures and trim nerve endings. Curved will dissect vessels and nerves. In addition to the tips, the sides may be used for dissection with the tips closed, similar to using forceps.