Multi-Cut Utility Scissors, OR Grade, Sklar
Supplier: Sklar
11-1278 11-1280 11-1281 11-1282 11-1283 11-1284 11-1286 11-1287
82029-880EA 61.4 CAD
82029-880 82029-882 89025-456 89025-458 89025-460 89025-462 10194-966 10194-968
Multi-Cut Utility Scissors, OR Grade, Sklar
Scissors
Utility Scissors are a multipurpose instrument used for cutting non-sterile materials such as dressings, casts, and surgical drapes.
  • Flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked
  • One larger plastic finger ring for maximized control while cutting
Scissors are angled with one serrated blade and one smooth blade. The flattened, wide, blunt tip on the serrated blade greatly reduces the likelihood of skin being nicked if the scissors are used for cutting material that is on the patient, such as clothing or bandages. They should never be used to cut tissue or sutures. One plastic finger ring is larger than the other for maximized control while cutting.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.