Stevens Tenotomy Scissors, OR Grade, Sklar
Supplier: Sklar
22-1240 64-1242 64-1342 64-1243 64-1343 64-1241 64-1341
89025-230EA 358.51 CAD
89025-230 10192-342 10192-360 10194-350 10194-352 10197-850 10197-856
Stevens Tenotomy Scissors, OR Grade, Sklar
Scissors
Stevens Tenotomy Scissors are used for delicate dissection and cutting in ophthalmologic, neurological, and plastic surgery procedures.
They have long handles and small, sharp blades with sharp or blunt wedge-shaped tips
Censitrac Ready OR Grade Stevens Tenotomy Scissors are industry standard and constructed of stainless steel.
The long handles allow the surgeon to have a firm grip and control while dissecting or cutting in small or narrow areas. Sharp tips allow precise dissection and cutting of delicate tissue, while blunt tipped blades are used more often for dissecting and cutting mucus membranes.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.