Starch 1% (w/v) in aqueous solution indicator, VWR Chemicals BDH®

Supplier: VWR International

BDH5104-1L BDH5104-4L
BDH5104-1LEA 80.62 CAD
BDH5104-1L BDH5104-4L
Starch 1% (w/v) in aqueous solution indicator, VWR Chemicals BDH®
Starch

This indicator is prepared using soluble starch and high purity water. Salicylic acid is added as a preservative. Store at 5 to 30 °C tightly closed in the original containers.

Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now

Specification Test Results

Appearance Translucent white liquid
Suitability for use Colorless (Iodine absent) - Blue (Iodine present)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR