Rochester-Carmalt Forceps, OR Grade, Sklar
Supplier: Sklar
17-1180 17-1162 17-1062 17-1080
10192-296EA 289.95 CAD
10192-296 10193-292 10194-732 10194-734
Rochester-Carmalt Forceps, OR Grade, Sklar
Forceps
Rochester-Carmalt Hemostatic Forceps are a standard type of hemostatic clamp used in several procedures.
The forceps have longitudinal serrations and longitudinal cross serrations. The fully serrated working surface allows this instrument to have greater grip on heavy or tough tissue to stop blood flow of large vessels. The ratcheted handle allows locking on larger tissue for hemostasis and constructed of stainless steel.Censitrac Ready products are industry standard.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.