Starch 0.5% (w/v) in aqueous solution, VWR Chemicals BDH®

Supplier: VWR International

BDH51020-1A BDH51020-500A
BDH5102-1LEA 112.5 CAD
BDH5102-1L BDH5102-500ML
Starch 0.5% (w/v) in aqueous solution, VWR Chemicals BDH®
Starch
Formula: (C₆H₁₀O₅)n
MW: 162.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00082026
CAS Number: 9005-25-8
EINECS: 232-679-6

Order Now

Specification Test Results

Appearance Translucent white liquid
Suitability for use Colorless (Iodine absent) - Blue (Iodine present)
Storage condition 5-30 °C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR