Jansen Bayonet Nasal Forceps, OR Grade, Sklar
Supplier: Sklar
70-4375 67-1465
10194-430EA 215.17 CAD
10194-430 10198-284
Jansen Bayonet Nasal Forceps, OR Grade, Sklar
Forceps
Sklar® Jansen Bayonet Dressing Forceps are thumb forceps commonly used in nasal procedures for placing packing.
The bayonet style handle and long jaws allow an unobstructed view when working in the nasal cavity. Serrated tips securely hold surgical materials. These forceps may also be used in intracranial procedures. This product has straight, serrated tips.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.