Carbon Film on London Finder Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
CFLF135-AU-50 CFLF400-AU-25 CFLF400-CU-25 CFLF135-CU-UL CFLF200-CU-UL CFLF400-CU-UL CFLF135-CU-TH CFLF200-CU-TH CFLF400-CU-TH CFLF135-CU-ET CFLF200-CU-ET CFLF400-CU-ET CFLF135-NI-UL CFLF200-NI-UL CFLF400-NI-UL CFLF135-AU-UL CFLF135-NI-TH CFLF200-NI-TH CFLF400-NI-TH CFLF135-NI-ET CFLF200-NI-ET CFLF400-NI-ET CFLF135-AU-TH CFLF135-AU-ET
102100-256BX 362.74 CAD
102100-256 102100-276 102100-278 103304-820 103304-822 103304-824 103304-898 103304-900 103304-902 103304-970 103304-972 103304-974 103305-506 103305-508 103305-510 103305-872 103306-014 103306-016 103306-018 103306-062 103306-064 103306-066 103306-122 103306-126
Carbon Film on London Finder Grids, Electron Microscopy Sciences
Graticules, Reticles and Grids
A thin film of pure carbon is deposited on the shiny side of the grid.
  • Carbon coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
  • Select copper, nickel, or gold grids
Optically checked followed by batch testing in the EM.
Packaging: Packed in grid storage box.