Noyes Alligator Forceps, OR Grade, Sklar
Supplier: Sklar
70-5335 70-5336 67-1734 70-5232 70-5235 70-5332
10193-310EA 1113.94 CAD
10193-310 10198-138 10198-286 10198-558 10198-560 10198-568
Noyes Alligator Forceps, OR Grade, Sklar
Forceps
Noyes Alligator Forceps are a finger ring instrument used for ENT procedures.
The long shaft allows access to deep, narrow spaces such as the ear and nasal canals. Serrated jaws provide additional grip for removing foreign objects. This product has straight 3" shanks that angle into a 5 1/2" shaft at the box lock. The tips are serrated with 1 x 2 teeth and a central groove as well.