Allis Tissue Forceps, OR Grade, Sklar
Supplier: Sklar
36-2060 22-6157 18-2164 36-1860 36-2295 18-2162 36-2280 18-2140 18-2155 36-1975 36-2380
82030-178EA 161.54 CAD
82030-178 89025-380 490010-486 490010-488 10192-334 10193-374 10194-086 10194-836 10194-838 10195-734 10195-738
Allis Tissue Forceps, OR Grade, Sklar
Forceps
Allis Tissue Forceps are finger ring, ratcheted forceps used for grasping organs and slippery or dense tissue
Censitrac Ready products are industry standard and constructed of stainless steel.
They are used with tonsil, vaginal, breast, and thyroid tissue.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.