Merit™ Rochester-Pean Forceps, Physician Grade, Sklar
Supplier: Sklar
97-470 97-471 97-477 97-481 97-483 97-485 97-487 97-492 97-494 97-496 97-498
10184-814EA 35.55 CAD
10184-814 10184-816 10184-818 10184-820 10184-822 10184-824 10184-826 10184-828 10184-830 10184-832 10184-834
Merit™ Rochester-Pean Forceps, Physician Grade, Sklar
Forceps
Merit™ Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis.
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight.
Merit™ instruments are mid-grade products used for routine and in-office procedures. These high quality instruments are crafted from Pakistan steel with a 10 year guarantee. Merit™ instruments deliver a solution for physicians and practices that require quality, durable instruments.