Surgi-OR™ Thumb Tissue Forceps, Mid Grade, Sklar
Supplier: Sklar
95-753 95-774
89023-702EA 31.95 CAD
89023-702 10184-628
Surgi-OR™ Thumb Tissue Forceps, Mid Grade, Sklar
Forceps
Surgi-OR™ Tissue Forceps are used for grasping, holding, and manipulating various types of tissues in multiple procedures.
These instruments have spring tension shanks, that determine how far the instrument will open and the thickness of the tissue that can be held. Some thumb tissue forceps have lesser tension and can only hold fine membranes or single layer tissue. Others have greater tension and can hold thick, heavy tissue. Thumb tissue forceps are commonly toothed in order to grip the tissue more firmly than serrated tips alone. They are available in several lengths to accommodate specific procedures, tissue types, and user preference.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel and provide reliable instrumentation for the operating room and other surgical centers.