Econo™ Sterile Kelly Hemostatic Forceps, Floor Grade, Sklar
Supplier: Sklar
96-2560 96-2561 96-2562 96-2563 96-2652 96-2653
89023-866CS 429.07 CAD
89023-866 89023-868 89023-870 89023-872 89023-962 89023-964
Econo™ Sterile Kelly Hemostatic Forceps, Floor Grade, Sklar
Forceps
Econo™ Sterile Kelly Hemostatic Forceps are used for clamping large blood vessels or manipulating heavy tissue. They may also be used for soft tissue dissection.
  • Ready-to-use
  • Available straight or curved
  • Ratcheted finger ring handle
The forceps are available curved or straight with a ratcheted finger ring handle
Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments.
The jaws of Kelly Forceps are 1/3 the length of the shanks and serrations are 1/2 the length of the jaws.