Econo™ Sterile Rochester-Pean Forceps, Sklar
Supplier: Sklar
96-2544 96-2545 96-2546 96-2547 96-2549 96-2664 96-2547A 96-3006 96-3008 96-3010
89023-834CS 1015.7 CAD
89023-834 89023-836 89023-838 89023-840 89023-842 89023-844 89023-986 10184-652 75995-438 75995-440 75995-442
Econo™ Sterile Rochester-Pean Forceps, Sklar
Forceps
Econo™ Sterile Rochester-Pean Hemostatic Forceps are used for clamping larger tissue and vessels for hemostasis
  • Full horizontal serrations
  • Versatile instrument used in multiple procedures
  • Available curved or straight
The full horizontal serrations and availability in multiple lengths make these forceps a versatile instrument used in multiple procedures. They are available curved or straight. Econo™ Sterile instruments are ready-to-use, sterile, disposable, floor-grade instruments made in Pakistan. Econo™ Sterile instruments eliminate the need for cleaning and reprocessing. Because they are both sterile and disposable, Econo™ Sterile instruments significantly reduce the likelihood of cross-contamination. They are available in bulk quantities.