Gerald Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
47-1175 47-1180
89024-944EA 222.89 CAD
89024-944 89024-946
Gerald Dressing Forceps, OR Grade, Sklar
Forceps
Sklar® Gerald Dressing Forceps are thumb style forceps commonly used for placing or removing dressings in many types of procedures.
Although Dressing Forceps were originally designed to specifically hold materials such as gauze, they have come to have a variety of additional functions
Censitrac Ready OR Grade Gerald Dressing Forceps are industry standard and constructed of stainless steel.
They may be used for removing foreign objects or dead tissue from wounds, as well as other small surgical materials such as tubing.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.