Innovating Science® Dialysis Tubings
Supplier: Ward's Science
ADDD0105-25 ADDD0106-100 ADDD0106-10 ADDD0107-100 ADDD0107-10 ADDD0108-100 ADDD0108-10 ADDD0108-50 ADDD0109-100 ADDD0109-10 ADDD0105-100 ADDD0105-10 ADDD0106500FT
470163-400EA 112.2 CAD
470163-400 470163-402 470163-404 470163-406 470163-408 470163-410 470163-412 470163-414 470163-416 470163-418 470163-424 470163-426 470231-460
Innovating Science® Dialysis Tubings
Tubing Dialysis Tubing
The semi-permeable membrane, created from a natural source of cellulose, allows passage of particles up to 14000 Da.
  • Available in multiple lengths
Flat width and open diameter measurement provided in the ordering table.