Surgi-OR™ Operating Scissors, Mid-Grade, Sklar
Supplier: Sklar
95-110 95-111 95-265 95-272 95-275 95-278 95-281 95-284 95-292 95-295 95-266
89023-452EA 308.07 CAD
89023-452 89023-454 89023-536 89023-538 89023-540 89023-542 89023-544 89023-546 89023-552 89023-554 10184-588
Surgi-OR™ Operating Scissors, Mid-Grade, Sklar
Scissors
Surgi-OR™ Operating scissors, known as surgical scissors are a versatile surgical instrument
With three available tip configurations, varying curvatures and multiple lengths, these scissors are used for general, multipurpose cutting and dissecting. They cut tougher tissue and are used for dissecting, whereas blunt Operating Scissors are used for cutting materials such as sutures, tubing, or gauze.
Hand-crafted in Pakistan, Surgi-OR™ mid-grade instruments are forged from high quality Japanese steel for the operating room and other surgical centers.
Order Now
SPECIFICATIONS
Environmentally Preferable