Micro Cover Glasses, Square, Matsunami Glass
Supplier: Matsunami Glass
C022221 C024241 C218181
10799-264BX 32.28 CAD
10799-264 10799-266 10026-134
Micro Cover Glasses, Square, Matsunami Glass
Cover Glasses
Matsunami Glass is a pioneer in the development of cover glass, and widely recognized as the standard for quality and flatness
Cover glass thickness rating: No 1 (0.12–0.17 mm).
Matsunami Micro Cover Glasses are manufactured from the highest grade of glass on a fully automated production line to ensure consistency.
Ordering information: Cover glasses are packaged 200 per box, 5 boxes per case (1000 items total).