Percoll® Density Gradient Media
Supplier: Cytiva
17-0891-02 17-0891-09 17-0891-01 17-0891-09 17-0891-01 17-0891-02 17-0891-02 17-0891-09 17-0891-01
CA11027-860EA 709 CAD
CA11027-860 CA11027-862 CA95021-202L CA95038-164L CA95038-166L CA11027-858 89428-526 89428-522 89428-524
Percoll® Density Gradient Media
Cell Separation Media
Percoll® is commonly used for separating cells, organelles, viruses, and other subcellular particles.
  • For research use only
  • Commonly used in basic research applications
  • Used in balanced salt solutions
  • Colorless to slightly yellow
A silica-based density gradient media for use in separating cells, organelles, viruses, and other subcellular particles. Percoll® comprises silica particles coated with polyvinylpyrrolidone (PVP) and is commonly used in basic research applications. Also used in balanced salt solutions, physiological saline or 0.25 M sucrose, and for formation of gradients either by the use of convenient gradient mixers or by high speed centrifugation. Colloidal solution of silica coated with polyvinylpyrrolidone (PVP). Liquid/suspension. Colorless to slightly yellow. Certificate of Analysis is available.
Caution: Percoll is for research use only.