Rxi®-5HT Columns, Restek
Supplier: Restek
13909 13910 13924 13906 13905 13908 13923
89217-490EA 1509.41 CAD
89217-490 89217-492 89217-502 89217-480 89217-474 89217-484 89217-496
Rxi®-5HT Columns, Restek
Chromatography Columns
These columns are ideal for GC/MS applications requiring high temperature resistance.
  • Columns processed for high-temperature applications
  • Low polarity
  • Up to 40% longer lifetime from specially designed fused silica tubing
Low-polarity phase, 5% diphenyl, 95% dimethyl polysiloxane. Temperature range: −60 to 400 °C (−76 to 752 °F). Similar phases: DB-5ht, VF-5ht, ZB-5HTinferno.