Mayo-Robson Intestinal Forceps, OR Grade, Sklar
Supplier: Sklar
36-3210 36-3310
10195-766EA 601.69 CAD
10195-766 10195-768
Mayo-Robson Intestinal Forceps, OR Grade, Sklar
Forceps
Mayo-Robson Intestinal Forceps are used for grasping or holding the bowel during intestinal procedures or anastomosis.
Longitudinal serrations grip the tissue firmly but gently, and the instrument is ratcheted for when locking the forceps is beneficial.