Anti-TRIB3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA515480
CAPIPA5-15480EA 3335.52 CAD
CAPIPA5-15480
Anti-TRIB3 Rabbit Polyclonal Antibody
Antibodies
NPK is a putative protein kinase that is induced by the transcription factor NF-kappaB. The encoded protein is a negative regulator of NF-kappaB and can also sensitize cells to TNF- and TRAIL-induced apoptosis. In addition, this protein can negatively regulate the cell survival serine-threonine kinase AKT1.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: TRIB3 (tribbles pseudokinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR