Anti-NTF3 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA514861
CAPIPA5-14861EA 3378.53 CAD
CAPIPA5-14861
Anti-NTF3 Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, chicken, porcine, rat and Xenopus based on sequence homology. NTF3 is a member of the neurotrophin family that controls survival and differentiation of mammalian neurons. This protein is closely related to both nerve growth factor and brain-derived neurotrophic factor. It may be involved in the maintenance of the adult nervous system, and may affect development of neurons in the embryo when it is expressed in human placenta. NTF3-deficient mice generated by gene targeting display severe movement defects of the limbs. The mature peptide of this protein is identical in all mammals examined including human, pig, rat and mouse.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: NTF3 (neurotrophin 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR