Anti-OMD Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA514382
CAPIPA5-14382EA 3258.01 CAD
CAPIPA5-14382
Anti-OMD Rabbit Polyclonal Antibody
Antibodies
OMD may be implicated in biomineralization processes. This protein has a function in binding of osteoblasts via the alpha(V)beta(3)-integrin.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry: 1:50-1:100, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: OMD (osteomodulin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR