Anti-BCL6 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA514259
CAPIPA5-14259EA 3694.23 CAD
CAPIPA5-14259
Anti-BCL6 Rabbit Polyclonal Antibody
Antibodies
BCL6 is a zinc finger transcription factor and contains an N-terminal POZ domain. This protein acts as a sequence-specific repressor of transcription, and has been shown to modulate the transcription of START-dependent IL-4 responses of B cells. This protein can interact with a variety of POZ-containing proteins that function as transcription corepressors.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:10-1:50, Western Blot: 1:1000

Type: Primary
Antigen: BCL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR