Anti-CKMM Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513830
CAPIPA5-13830EA 3406.91 CAD
CAPIPA5-13830
Anti-CKMM Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, porcine and rabbit based on sequence homology. Creatine kinase isoenzymes play a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa. The CKM isoform, predominant in skeletal muscle and heart tissue, is a cytoplasmic enzyme involved in energy homeostasis and is an important serum marker for myocardial infarction. CKM reversibly catalyzes the transfer of phosphate between ATP and various phosphogens such as creatine phosphate. It acts as a homodimer in striated muscle as well as in other tissues, and as a heterodimer with a similar brain isozyme in heart. The encoded protein is a member of the ATP:guanido phosphotransferase protein family.

Flow Cytometry: 1:10-1:50, Immunohistochemistry: 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: CKMM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR