Anti-ETV4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513595
CAPIPA5-13595EA 3335.52 CAD
CAPIPA5-13595
Anti-ETV4 Rabbit Polyclonal Antibody
Antibodies
ETV4 is a transcriptional activator that binds to the enhancer of the adenovirus E1A gene; the core-binding sequence is 5'[AC]GGA[AT]GT-3'.

Recommended Dilutions: Western Blot: 1:1000, Immunohistochemistry (Paraffin): 1:10 - 1:50, Flow Cytometry: 1:10-1:50

Type: Primary
Antigen: ETV4 (ets variant 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR