Anti-PRKRA Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA513022
CAPIPA5-13022EA 3258.01 CAD
CAPIPA5-13022
Anti-PRKRA Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine and rat based on sequence homology. PRKRA appears to have a pro-apoptotic function that may be suppressed in the presence of growth factor. It activates EIF2AK2 in the absence of double-stranded RNA (dsRNA).

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: PRKRA (protein kinase, interferon-inducible double stranded RNA dependent activator)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR