Anti-MAP4K4 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512858
CAPIPA5-12858EA 3378.53 CAD
CAPIPA5-12858
Anti-MAP4K4 Rabbit Polyclonal Antibody
Antibodies
M4K4, a member of the STE20 subfamily of Ser/Thr protein kinases, may play a role in the response to environmental stress and cytokines such as TNF-alpha. It appears to act upstream of the JUN N-terminal pathway. This protein is thought to interact with the SH3 domain of the adapter proteins Nck. HGK binds, via its CNH regulatory domain, to the N-terminal region of SPG3A. Expression appears to be ubiquitous, expressed in all tissue types examined. Isoform 5 appears to be more abundant in the brain, and isoform 4 is predominant in the liver, skeletal muscle and placenta.

Recommended Dilutions: Dot blot: 1:500

Type: Primary
Antigen: MAP4K4 (mitogen-activated protein kinase kinase kinase kinase 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR