Anti-ALCAM Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512513
CAPIPA5-12513EA 3485.16 CAD
CAPIPA5-12513
Anti-ALCAM Rabbit Polyclonal Antibody
Antibodies
This antibody is predicted to react with bovine, mouse, rat and rabbit based on sequence homology. CD166 is cell adhesion molecule that binds to CD6. The protein is involved in neurite extension by neurons via heterophilic and homophilic interactions. It may play a role in the binding of T- and B-cells to activated leukocytes, as well as in interactions between cells of the nervous system.

Flow Cytometry: 1:10-1:50, Immunohistochemistry (Paraffin): 1:50-1:100, Western Blot: 1:1000

Type: Primary
Antigen: ALCAM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR