Anti-EXO1 Rabbit Polyclonal Antibody

Supplier: Pierce Biotechnology
PA512501
CAPIPA5-12501EA 3335.52 CAD
CAPIPA5-12501
Anti-EXO1 Rabbit Polyclonal Antibody
Antibodies
EXO1 is a protein with 5' to 3' exonuclease activity as well as an RNase H activity. This protein is similar to the Saccharomyces cerevisiae protein Exo1 which interacts with Msh2 and which is involved in mismatch repair and recombination.

Recommended Dilutions: Western Blot: 1:1000, Immunofluorescence: 1:10-1:50, Immunocytochemistry: 1:10-1:50

Type: Primary
Antigen: EXO1 (exonuclease 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR