Anti-SSEA5 Monoclonal Antibody

Supplier: Pierce Biotechnology
MA1144D488
CAPIMA1144D488EA 2126.93 CAD
CAPIMA1144D488
Anti-SSEA5 Monoclonal Antibody
Antibodies
Immunofluorescence and flow cytometric analysis of MA1-144-D488 shows positive staining of SSEA-5 positive pluripotent cells and cultured human embryonal carcinoma NCCIT cells. Stage-Specific Embryonic Antigen-5 (SSEA-5) is an oligosaccharide that can be part of glycosphingolipids and glycoproteins. It contains the motif Fuc1-2Gal-beta1-3GlcNAc-beta, which is characteristic of H-1 antigens. SSEA-5 is found in the inner cell mass of the human blastocyst during embryogenesis, on undifferentiated human embryonic stem cells and on the surface of induced pluripotent cells. SSEA-5 was found to proceed the loss of other known stem cell markers and the combination of SSEA-5 with either CD9 and CD90 or CD50 and CD200 was superior in identifying and eliminating rare undifferentiated human ES cells from differentiated cell populations than the use of other established stem cell markers.

Recommended Dilutions: Immunofluorescence: 1:10-1:100, Immunocytochemistry: 1:10-1:100, Flow Cytometry: 1:50-1:500

Type: Primary
Antigen: SSEA5
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host:
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR