It has been successfully used in ICC/IF and flow cytometry applications on human samples. TRA-1-81 is a cell surface antigen expressed along with SSEA-3, SSEA-4 and TRA-1-60 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are down-regulated during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
This Antibody has been successfully used in ICC/IF and flow cytometry applications on human samples. TRA-1-81 is a cell surface antigen expressed along with SSEA-3, SSEA-4 and TRA-1-60 in human embryonic stem cells, embryonal carcinoma cells and induced pluripotent stem cells (iPS). These surface markers are down-regulated during the differentiation process. In contrast, SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.
Type: Primary
Antigen: TRA-1-81
Clonality: Monoclonal
Clone: tra-1-81
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human