Anti-CD15 Mouse Monoclonal Antibody (DyLight® 550) [clone: MC-480]

Supplier: Pierce Biotechnology
MA1022D550
CAPIMA1022D550EA 1989.65 CAD
CAPIMA1022D550
Anti-CD15 Mouse Monoclonal Antibody (DyLight® 550) [clone: MC-480]
Antibodies
It has been successfully used in flow cytometry on mouse and human samples. SSEA-1 (stage-specific embryonic antigen-1) is a cell surface antigen, expressed in murine embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.

This Antibody has been successfully used in flow cytometry on mouse and human samples. SSEA-1 (stage-specific embryonic antigen-1) is a cell surface antigen, expressed in murine embryonic stem cells, embryonal carcinoma cells, and induced pluripotent stem cells (iPS). SSEA-1 is absent in undifferentiated human stem cells but is present on the cell surface after retinoic acid mediated differentiation.

Type: Primary
Antigen: CD15
Clonality: Monoclonal
Clone: MC-480
Conjugation: DyLight® 550
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR