Sklarlite™ Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
23-2824 23-2826 23-2828 23-2830
10195-354EA 89.67 CAD
10195-354 10195-356 10195-358 10195-360
Sklarlite™ Dressing Forceps, OR Grade, Sklar
Forceps
Sklarlite™ Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
Sklarlite™ instruments are a lighter version of standard Sklar instruments. A unique manufacturing process developed by Sklar shaves microns off standard and delicate pattern instruments while maintaining structural integrity and durability. This lightweight modification provides the surgeon with greater control and less fatigue.
Order Now
SPECIFICATIONS
Environmentally Preferable