Cartesian Diver Set
Supplier: Shiv Dial Sud & Sons
PH10634 PH10634/D
470218-908CS 627.95 CAD
470218-908 470218-910
Cartesian Diver Set
Educational Materials Physics Educational Materials Physics Fundamentals Learning Activities
This diver set expresses the effects of air pressure and buoyancy.
  • Topics Include Transmission of Pressure by Liquid, Compressibility of Gases, and Archimedes’ Principle
  • Set Also Expresses Effects of Air Pressure and Buoyancy
  • Instructions Included
This apparatus is ideal for showing the transmission of pressure by a liquid, the compressibility of gases, and Archimedes’ Principle.
Instructions are included.
Ordering information: This kit includes a Cartesian diver, a 25 x 4 cm dia. glass hydrometer cylinder, a rubber diaphragm, and instructions.