UltraPlex 1-Step ToughMix (4X), QuantaBio
Supplier: Quantabio
95166-100 95166-500 95167-100 95168-100 95168-500 95166-01K 95166-01K 95166-100 95166-500 95167-100 95168-100 95168-500 95166-10K 95168-10K
10804-944EA 464.52 CAD
10804-944 10804-946 10804-948 10804-950 10804-952 10804-954 CA76102-178 76121-382 CA10804-944 CA10804-946 CA10804-948 CA10804-952 CA10804-954 CA76518-322 CA76518-324
UltraPlex 1-Step ToughMix (4X), QuantaBio
Nucleic Acid Reagents qPCR/RT-qPCR Enzymes and Kits
UltraPlex 1-Step ToughMix is a ready-to-use, 4X concentrated master mix 1-step reverse transcription and real-time quantitative PCR (RT-qPCR) of RNA templates using probe-based detection methods. First-strand cDNA synthesis and PCR amplification are carried out in the same tube without opening between steps. Optimized to deliver highly sensitive quantification of RNA viruses or low abundance RNA targets in uni- or highly multiplexed RNA detection assays, this reagent chemistry is optimized to deliver maximum assay sensitivity, precision and reproducibility with miniaturized reaction volumes and either conventional or accelerated thermal cycling protocols. UltraPlex 1-Step ToughMix contains all required components for RT-qPCR except RNA template and probe and is compatible with all dual-labeled probe chemistries.
  • 4X concentrated master mix allows greater flexibility with diluted samples (as little as 1 pg of total RNA) and multiplexing up to 5 targets
  • Tough-tested for applications where PCR inhibition from template impurities may be a concern
  • Temperature stabilized master mix enables convenient reaction setup at room temperature
UltraPlex 1-Step ToughMix (Low ROX/ ROX) (4X), contains:
4X reaction buffer containing optimized titrations of qPCR-grade dATP, dCTP, dGTP, dTTP, and magnesium.
Ultra-pure recombinant proteins: qScript XLT reverse transcriptase, RNase inhibitor protein, and AccuStart II hot-start Taq DNA polymerase.
Proprietary stabilizers additives and optimized ROX passive reference dye (if applicable).
The system has been optimized to deliver maximum RT‑qPCR efficiency, sensitivity, and specificity in reduced reaction volumes and fast cycle times. UltraPlex 1-Step ToughMix contains all required components for RT-qPCR except RNA template and probe. It is compatible with all dual-labeled probe chemistries.
Caution: UltraPlex 1-Step ToughMix is intended for molecular biology applications. This product is not intended for the diagnosis, prevention or treatment of a disease. Store components in a constant temperature freezer at -25°C to -15°C protected from light upon receipt.