Biacore consumables and accessories, Cytiva
Supplier: Cytiva
BR100541 BR100838 BR100286 BR100691 28953828 BR100058 BR100539 BR100556 BR100557 29-1496-03 29207950
101724-672EA 221.58 CAD
101724-672 101724-704 101724-792 101724-624 101724-736 101724-766 101725-264 101724-602 101724-700 101724-722 101724-724 102985-680 103568-390
Biacore consumables and accessories, Cytiva
Protein Purification Protein Purification Equipment
Biacore maintenance kits are convenient kits for proper instrument maintenance.
*Not recommended for use with Biacore 4000, Biacore A100 or Biacore S51.
Ordering information: The use of the CAPture kits in Biacore systems requires an Amine Coupling Kit with Sensor Chip CM5 CM4 CM3 or C. Not for use with Biacore Flexchip unless otherwise noted.
Order Now
SPECIFICATIONS
Environmentally Preferable