Mouse Recombinant VEGF (from E. coli)
Supplier: Peprotech
AF-450-32-2UG AF-450-32-10UG AF-450-32-100UG AF-450-32-250UG AF-450-32-500UG AF-450-32-1MG
10781-846EA 289.23 CAD
10781-846 10781-848 10781-850 10781-852 10781-854 10781-856
Mouse Recombinant VEGF (from E. coli)
Proteins and Peptides
VEGF is a potent growth and angiogenic cytokine. It stimulates proliferation and survival of endothelial cells, and promotes angiogenesis and vascular permeability. Expressed in vascularized tissues, VEGF plays a prominent role in normal and pathological angiogenesis. Substantial evidence implicates VEGF in the induction of tumor metastasis and intra-ocular neovascular syndromes. VEGF signals through three receptors; fms-like tyrosine kinase (flt-1), KDR gene product (the murine homolog of KDR is the flk-1 gene product) and the flt4 gene product. Recombinant Murine VEGF
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.