Human Recombinant NT4 (from E. coli)
Supplier: Peprotech
AF-450-04-2UG AF-450-04-10UG AF-450-04-100UG AF-450-04-250UG AF-450-04-500UG AF-450-04-1MG
10781-808EA 304.45 CAD
10781-808 10781-810 10781-812 10781-814 10781-816 10781-818
Human Recombinant NT4 (from E. coli)
Proteins and Peptides
NT-4 is a neurotrophic factor structurally related to β-NGF, BDNF, and NT-3.  These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures.  NT-4 is expressed in the prostate, thymus, placenta, and skeletal muscle.  NT-4 can signal through the LNGFR and trkB receptors, and promotes the survival of peripheral sensory sympathetic neurons.  Recombinant Human NT-4 is a noncovalently linked homodimer of two 14.0 kDa polypeptide monomers (260 total amino acid residues). Manufactured using all Animal-Free reagents.
  • Carrier free
  • Animal-free
GVSETAPASR RGELAVCDAV SGWVTDRRTA VDLRGREVEV LGEVPAAGGS PLRQYFFETR CKADNAEEGG PGAGGGGCRG VDRRHWVSEC KAKQSYVRAL TADAQGRVGW RWIRIDTACV CTLLSRTGRA
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.