Human Recombinant IL10 (from E. coli)
Supplier: Peprotech
AF-200-10-100UG AF-200-10-250UG AF-200-10-500UG AF-200-10-1MG AF-200-10-2UG AF-200-10-10UG AF-200-10-50UG
10771-712EA 3430.76 CAD
10771-712 10771-714 10771-716 10771-718 10781-754 10781-756 10781-758
Human Recombinant IL10 (from E. coli)
Proteins and Peptides
IL-10 is an immunosuppressive cytokine produced by a variety of mammalian cell types including macrophages, monocytes, T cells, B cells and keratinocytes.  IL-10 inhibits the expression of proinflammatory cytokines such as IL-1 and TNF-α.  Like IL-4, IL-10 enhances humoral immune responses and attenuates cell-mediated immune reactions.  Human IL-10 is active on murine cells, but murine IL-10 is inactive on human cells.  Recombinant Human IL-10 is an 18.6 kDa protein of 161 amino acid residues. Manufactured using all Animal-Free reagents.
  • Carrier free
  • Animal-free
MSPGQGTQSE NSCTHFPGNL PNMLRDLRDA FSRVKTFFQM KDQLDNLLLK ESLLEDFKGY LGCQALSEMI QFYLEEVMPQ AENQDPDIKA HVNSLGENLK TLRLRLRRCH RFLPCENKSK AVEQVKNAFN KLQEKGIYKA MSEFDIFINY IEAYMTMKIR N
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.