Human Recombinant IL9 (from E. coli)
Supplier: Peprotech
AF-200-09-2UG AF-200-09-10UG AF-200-09-50UG AF-200-09-100UG AF-200-09-250UG AF-200-09-500UG AF-200-09-1MG
10781-740EA 289.23 CAD
10781-740 10781-742 10781-744 10781-746 10781-748 10781-750 10781-752
Human Recombinant IL9 (from E. coli)
Proteins and Peptides
IL-9 is an immunoregulatory cytokine produced by IL-2 activated Th2 lymphocytes.  IL-9 enhances the proliferation of T lymphocytes, mast cells, erythroid precursor cells and megakaryoblastic leukemia cell lines.  Over-expression of IL-9 has been implicated in the pathogenesis of anaplastic lymphoma and Hodgkin’s disease.  Whereas murine IL-9 can function on human cells, human IL-9 is inactive on mouse cells.  Recombinant Human IL-9 is a 14.0 kDa protein of 127 amino acid residues, including 10 cysteine residues that are fully conserved between the human murine proteins. Manufactured using all Animal-Free reagents.
  • Carrier free
  • Animal-free
MQGCPTLAGI LDINFLINKM QEDPASKCHC SANVTSCLCL GIPSDNCTRP CFSERLSQMT NTTMQTRYPL IFSRVKKSVE VLKNNKCPYF SCEQPCNQTT AGNALTFLKS LLEIFQKEKM RGMRGKI
Verified by N-terminal and Mass Spectrometry analyses (when applicable).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.