Human Recombinant FGF-acidic (from E. coli)
Supplier: Peprotech
AF-100-17A-10UG AF-100-17A-50UG AF-100-17A-100UG AF-100-17A-250UG AF-100-17A-500UG AF-100-17A-1MG
10781-708EA 289.23 CAD
10781-708 10781-710 10781-712 10781-714 10781-716 10781-718
Human Recombinant FGF-acidic (from E. coli)
Proteins and Peptides
FGF-acidic is one of 23 known members of the FGF family. Proteins of this family play a central role during prenatal development, postnatal growth and regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-acidic is a non-glycosylated heparin binding growth factor that is expressed in the brain, kidney, retina, smooth muscle cells, bone matrix, osteoblasts, astrocytes and endothelial cells. FGF-acidic has the ability to signal through all the FGF receptors. Recombinant Human FGF-acidic is a 16.8 kDa protein consisting of 141 amino acid residues.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.