Mouse Recombinant FGF-23 (from E. coli)
Supplier: Peprotech
450-55-5UG 450-55-20UG 450-55-100UG 450-55-250UG 450-55-500UG
10781-442EA 239.65 CAD
10781-442 10781-444 10781-446 10781-448 10781-450
Mouse Recombinant FGF-23 (from E. coli)
Proteins and Peptides
The FGF family plays a central role during prenatal development, postnatal growth, and the regeneration of a variety of tissues, by promoting cellular proliferation and differentiation. FGF-23, FGF-21 and FGF-19 constitute an atypical FGF subfamily whose ligands act as circulating hormones and require the participation of a Klotho protein as a co-receptor for their signaling. FGF-23 is a bone-derived hormone that acts in the kidney to regulate phosphate homeostasis and vitamin D metabolism. The signaling receptor for FGF-23, a Klotho-FGFR1 (IIIc) complex, is an essential regulator of the renal sodium phosphate co-transporter and key vitamin D-metabolizing enzymes CYP27B1 and CYP24A1. Recombinant Murine FGF-23 is a 25.5 kDa globular protein containing 228 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.