Lysobacter Recombinant Enzymogenes (from Hi-5 Insect Cells)
Supplier: Peprotech
450-54-5UG 450-54-20UG 450-54-100UG
10781-430EA 209.84 CAD
10781-430 10781-432 10781-434 10781-436 10781-438 10781-440
Lysobacter Recombinant Enzymogenes (from Hi-5 Insect Cells)
Proteins and Peptides
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Arg-C specifically cleaves at the carboxyl side of Arginine residues. Arg-C has a sulfhydryl requirement; it is activated by dithiothreitol, cysteine, or other sulfhydryl-containing reagents. The presence of calcium ions is essential. The enzyme is inhibited by oxidizing agents and sulfhydryl reactants, and by Co2+, Cu2+, Cd2+, and heavy metal ions. Recombinant Lysobacter Enzymogenes Arg-C is a 26.8 kDa protease consisting of 252 amino acid residues including a C-terminal His-Tag.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.