Human Recombinant Enterokinase (from CHO cells)
Supplier: Peprotech
450-48C-10UG 450-48C-50UG 450-48C-100UG 450-48C-250UG 450-48C-500UG 450-48C-1MG
10781-392EA 209.84 CAD
10781-392 10781-394 10781-396 10781-398 10781-400 10781-402
Human Recombinant Enterokinase (from CHO cells)
Enzymes
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to, specific residues, or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Enterokinase sequentially cleaves carboxyl side of D-D-D-D-K. Human Enterokinase is expressed as a linear 1019 amino acid polypeptide precursor glycoprotein. Proteolytic processing of this precursor generates the biologically active form of Enterokinase, which consists of two polypeptide chains (heavy chain and light chain) held together by a single disulfide bond, resulting in formation of a biologically active heterodimer. The heavy chain consists of 784 amino acid residues, and the light chain consists of 235 amino acid residues. The calculated molecular weight of Recombinant Human Enterokinase is 108.7 kDa.
  • Origin: USA
  • Accession number: P98073
  • Gene ID: 5651
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.