Human Recombinant Furin (from Hi-5 Insect Cells)
Supplier: Peprotech
450-47-2UG 450-47-10UG 450-47-100UG 450-47-250UG
10781-380EA 209.84 CAD
10781-380 10781-382 10781-384 10781-386 10781-388 10781-390
Human Recombinant Furin (from Hi-5 Insect Cells)
Proteins and Peptides
Proteases (also called Proteolytic Enzymes, Peptidases, or Proteinases) are enzymes that hydrolyze the amide bonds within proteins or peptides. Most proteases act in a specific manner, hydrolyzing bonds at, or adjacent to specific residues or a specific sequence of residues contained within the substrate protein or peptide. Proteases play an important role in most diseases and biological processes, including prenatal and postnatal development, reproduction, signal transduction, the immune response, various autoimmune and degenerative diseases, and cancer. They are also an important research tool, frequently used in the analysis and production of proteins. Furin is a calcium-dependent serine endoprotease that processes numerous proproteins of different secretory pathways into their mature forms by cleaving at the carboxyl side of the recognition sequence, R-Xaa-(K/R)-R, where Xaa can be any amino acid. Recombinant Human Furin is a 63.9 kDa protein, corresponding to residues 131 through 715 of the Furin precursor, plus a C-terminal His-tag.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.